Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ANG-1 Recombinant Protein, PeproTech®

Catalog No. 1300620UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 100 μg
100 μg
20 μg
5 μg
5 x 100 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-06-20UG 20 μg
13006100UG 100 μg
13-006-500 5 x 100 μg
13-006-01M 10 x 100 μg
130-06-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 130-06-20UG Supplier Gibco™ Supplier No. 1300620UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

130-06-1MG will be provided as 10 x 100 μg (130-06-100UG). 130-06-500UG will be provided as 5 x 100 μg (130-06-100UG). Recombinant Human ANG-1, derived from HeLa cells, is a C-terminal histidine tagged glycoprotein which migrates with an apparent molecular mass of 60.0-70.0 kDa by SDS-PAGE under reducing conditions. Sequencing analysis shows N-terminal sequences starting with Ser-20 and with Asp-70 of the 498 amino acid precursor protein. The calculated molecular weight of Recombinant Human ANG-1 is 56.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15389
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 284
Molecular Weight (g/mol) 56.3 kDa
Name Human ANG-1
Purification Method Purified
Quantity 20 μg
Source HeLa cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188
Common Name Angiopoietin 1
Gene Symbol Angpt1
Biological Activity Determined by its ability to induce adhesion of human umbilical vein endothelial cells (HUVEC).
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence SNQRRSPENS GRRYNRIQHG QCAYTFILPE HDGNCRESTT DQYNTNALQR DAPHVEPDFS SQKLQHLEHV MENYTQWLQK LENYIVENMK SEMAQIQQNA VQNHTATMLE IGTSLLSQTA EQTRKLTDVE TQVLNQTSRL EIQLLENSLS TYKLEKQLLQ QTNEILKIHE KNSLLEHKIL EMEGKHKEEL DTLKEEKENL QGLVTRQTYI IQELEKQLNR ATTNNSVLQK QQLELMDTVH NLVNLCTKEG VLLKGGKREE EKPFRDCADV YQAGFNKSGI YTIYINNMPE PKKVFCNMDV NGGGWTVIQH REDGSLDFQR GWKEYKMGFG NPSGEYWLGN EFIFAITSQR QYMLRIELMD WEGNRAYSQY DRFHIGNEKQ NYRLYLKGHT GTAGKQSSLI LHGADFSTKD ADNDNCMCKC ALMLTGGWWF DACGPSNLNG MFYTAGQNHG KLNGIKWHYF KGPSYSLRST TMMIRPLDFH HHHHH
Content And Storage -20°C
Activity Determined by its ability to induce adhesion of human umbilical vein endothelial cells (HUVEC).
Expression System HeLa cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.