Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ANG-2 Recombinant Protein, PeproTech®

Catalog No. 13007250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13007250UG 250 μg
13007100UG 100 μg
130-07-1MG 1 mg
130-07-20UG 20 μg
13007500UG 500 μg
130-07-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 13007250UG Supplier Gibco™ Supplier No. 13007250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human ANG-2 is a C-terminal histidine-tagged glycoprotein which migrates with an apparent molecular mass of 60.0-70.0 kDa by SDS-PAGE under reducing conditions. Sequencing analysis shows an N-terminal sequence starting with residue 68 (D) of the ANG-2 precursor protein. The calculated molecular weight of Recombinant Human ANG-2 is 50.1 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

ANG-2 binds to the endothelial cell specific receptor Tie-2, but, in contrast to ANG-1, does not induce tyrosine phosphorylation. Consequently, ANG-2 modulates ANG-1 activation of Tie-2 and, depending on the physiological and biochemical environment, can act either as an agonist or antagonist of Tie-2 induced angiogenesis. The signaling interactions of ANG-1, ANG-2 and Tie-2, along with less characterized ANG-3 and ANG-4, are required for embryonic and adult angiogenesis. Physiologically, ANG-1 and ANG-2 are associated with sprouting, tube formation, and structural integrity of newly formed blood vessels. Mature human ANG-2 is a secreted protein containing 480 amino acid residues. ANG-2 is composed of an alpha-helix-rich coiled coil N-terminal domain and fibrinogen-like C-terminal domain. ANG-2 exists predominantly in the form of a disulfide-linked dimer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15123
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 285
Molecular Weight (g/mol) 50.1 kDa
Name Human ANG-2
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AGPT2; Ang2; Ang-2; angiopoietin 2; angiopoietin-2; angiopoietin-2 A; angiopoietin-2 B; ANGPT2; endothelial growth factor; Tie2-ligand
Common Name Angiopoietin 2
Gene Symbol ANGPT2
Biological Activity Determined by its ability to stimulate tubulogenesis in HUVEC cells using a concentration of 0.2 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence DAPLEYDDSV QRLQVLENIM ENNTQWLMKL ENYIQDNMKK EMVEIQQNAV QNQTAVMIEI GTNLLNQTAE QTRKLTDVEA QVLNQTTRLE LQLLEHSLST NKLEKQILDQ TSEINKLQDK NSFLEKKVLA MEDKHIIQLQ SIKEEKDQLQ VLVSKQNSII EELEKKIVTA TVNNSVLQKQ QHDLMETVNN LLTMMSTSNS AKDPTVAKEE QISFRDCAEV FKSGHTTNGI YTLTFPNSTE EIKAYCDMEA GGGGWTIIQR REDGSVDFQR TWKEYKVGFG NPSGEYWLGN EFVSQLTNQQ RYVLKIHLKD WEGNEAYSLY EHFYLSSEEL NYRIHLKGLT GTAGKISSIS QPGNDFSTKD GDNDKCICKC SQMLTGGWWF DACGPSNLNG MYYPQRQNTN KFNGIKWYYW KGSGYSLKAT TMMIRPADFH HHHHH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.