Learn More
ACROBiosystems Human Angiopoietin-like 3 1mg
Supplier:  ACROBiosystems AN3H52531MG
Encompass_Preferred
Human Angiopoietin-like 3 Protein Fc Tag (AN3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Glu 460 (Accession # Q9Y5C1-1).
Catalog No. 50-253-5506
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.