Learn More
Abcam Human Angiotensin II ELISA Kit, 96TEST
Supplier:  Abcam AB28530696TEST
Encompass_Preferred
Human Angiotensin II ELISA Kit is a Competitive ELISA kit for the measurement of Human Angiotensin II in Human in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2282109
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.