Learn More
Sino Biological Angiopoietin-1 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 13667MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Angiopoietin-1 (rh Angiopoietin-1; Catalog#13667-H02H1; NP_001137.2; Met1-Leu261 and Arg277-Phe498). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4195
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.