Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AOC2 Full-length ORF (NP_033720.2, 1 a.a. - 756 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89935181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-181 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-181 Supplier Abnova™ Supplier No. H00000314P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes and ammonia in the presence of copper and quinone cofactor. This gene shows high sequence similarity to copper amine oxidases from various species ranging from bacteria to mammals. The protein contains several conserved motifs including the active site of amine oxidases and the histidine residues that likely bind copper. It may be a critical modulator of signal transmission in retina, possibly by degrading the biogenic amines dopamine, histamine, and putrescine. This gene may be a candidate gene for hereditary ocular diseases. Alternate splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MHLKIVLAFLALSLITIFALAYVLLTSPGGSSQPPHCPSVSHRAQPWPHPGQSQLFADLSREELTAVMRFLTQRLGPGLVDAAQAQPSDNCIFSVELQLPPKAAALAHLDRGSPPPAREALAIVLFGGQPQPNVSELVVGPLPHPSYMRDVTVERHGGPLPYHRRPVLRAEFTQMWRHLKEVELPKAPIFLSSTFNYNGSTLAAVHATPRGLRSGDRATWMALYHNISGVGLFLHPVGLELLLDHRALDPAHWTVQQVFYLGHYYADLGQLEREFKSGRLEVVRVPLPPPNGASSLRSRNSPGPLPPLQFSPQGSQYSVQGNLVVSSLWSFTFGHGVFSGLRIFDVRFQGERIAYEVSVQECVSIYGADSPKTMLTRYLDSSFGLGRNSRGLVRGVDCPYQATMVDIHILVGKGAVQLLPGAVCVFEEAQGLPLRRHHNYLQNHFYGGLASSALVVRSVSSVGNYDYIWDFVLYPNGALEGRVHATGYINTAFLKGGEEGLLFGNRVGERVLGTVHTHAFHFKLDLDVAGLKNWVVAEDVVFKPVAAPWNPEHWLQRPQLTRQVLGKEDLTAFSLGSPLPRYLYLASNQTNAWGHQRGYRIQIHSPLGIHIPLESDMERALSWGRYQLVVTQRKEEESQSSSIYHQNDIWTPTVTFADFINNETLLGEDLVAWVTASFLHIPHAEDIPNTVTLGNRVGFLLRPYNFFDEDPSIFSPGSVYFEKGQDAGLCSINPVACLPDLAACVPDLPPFSYHGF

Specifications

Accession Number NP_033720.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 314
Molecular Weight (g/mol) 110.1kDa
Name AOC2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DAO2/RAO
Common Name AOC2
Gene Symbol AOC2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.