Learn More
Sino Biological APLP1 / Amyloid-like protein 1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 13716T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human APLP1 / Amyloid-like protein 1 (rh APLP1 / Amyloid-like protein 1; Catalog#13716-H08H; P51693-1; Met1-Glu580). APLP1 / Amyloid-like protein 1 specific IgG was purified by Human APLP1 / Amyloid-like protein 1 affinity chromatography.
Catalog No. 50-255-4193
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.