Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human APO A1, Apolipoprotein A-1 ELISA Kit

Catalog No. MP08L100039 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100039 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100039 Supplier MP Biomedicals™ Supplier No. 08L100039
Add to Cart
Add to Cart
This item is not returnable. View return policy

Quantitative detection of human APOA1/Apolipoprotein A1 in samples of cell culture supernatant, saliva, urine, serum, plasma, cell lysates, tissue homogenates, and other biological fluids.

Human APOA1/Apolipoprotein A1 protein participates in the reverse transport of cholesterol from tissues to the liver for excretion by promoting cholesterol efflux from tissues and by acting as a cofactor for the lecithin cholesterol acyltransferase. Defects in APOA1/Apolipoprotein A 1 are a cause of familial hypoalphalipoproteinemia and the low HDL levels as well as amyloidosis van Allen type disease.

  • In this ELISA kit, an APOA1/Apolipoprotein A1 specific antibody has been pre-coated onto 96-well plates and blocked
  • Standards or test samples are added to the wells and subsequently an APOA1/Apolipoprotein A1 specific biotinylated detection antibody is added and followed by washing with wash buffer
  • Afterwards, Streptavidin-Peroxidase Complex is added and unbound conjugates are washed away with wash buffer
  • TMB is then used to visualize Streptavidin-Peroxidase enzymatic reaction
  • TMB is catalyzed by Streptavidin-Peroxidase to produce a blue color product that changes to yellow after adding an acidic stop solution
  • The density of yellow coloration is directly proportional to the amount of APOA1/Apolipoprotein A1 captured in the plate
  • Sensitivity: 0.188 ng/mL in ELISA assays
TRUSTED_SUSTAINABILITY

Specifications

Assay Sandwich Assay
Conjugate APO A1
Cross Reactivity None
Immunoassay Kit Format 96-well Plate
Label Type Biotin
Product Line MP Biomedicals
Product Type ELISA Kit
Protein Family Apolipoprotein
Sample Type Plasma, Serum, Tissue Homogenates
Specificity No Cross-reactivity to Other Antigens
Substrate TMB
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
Sufficient For 96-well Plate
Target Species Human
Quantity 96 Preps
Regulatory Status RUO
Storage Requirements 2°C to 8°C
Detection Limit 0.313 ng/mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.