Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Apoe2 (Np_000032.1, 300 Amino Acids) Partial Recombinant Protein Expressed In Escherichia Coli.

Catalog No. 89035656 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-656 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-656 Supplier Abnova™ Supplier No. P6209
High quality

Specifications

Accession Number NP_000032.1
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 348
Name APOE2 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 50 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AD2/LPG/MGC1571/apoprotein
Common Name APOE
Gene Symbol APOE
Species E. coli
Protein Tag None
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.