Learn More
Immunology Consultants Human Apolipoprotein H (Beta-2-Glycoprotein) ELISA Kit
SDP
Supplier:  Immunology Consultants E80GLY
Encompass_Preferred
The Human Apo-H test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring Apo-H in human biological samples.
Catalog No. 50-240-6607
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.