Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human APOL6 Full-length ORF (NP_085144.1, 1 a.a. - 343 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89935384 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-384 25 μg
89-935-383 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-935-384 Supplier Abnova™ Supplier No. H00080830P01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. [provided by RefSeq]

Sequence: MDNQAERESEAGVGLQRDEDDAPLCEDVELQDGDLSPEEKIFLREFPRLKEDLKGNIDKLRALADDIDKTHKKFTKANMVATSTAVISGVMSLLGLALAPATGGGSLLLSTAGQGLATAAGVTSIVSGTLERSKNKEAQARAEDILPTYDQEDREDEEEKADYVTAAGKIIYNLRNTLKYAKKNVRAFWKLRANPRLANATKRLLTTGQVSSRSRVQVQKAFAGTTLAMTKNARVLGGVMSAFSLGYDLATLSKEWKHLKEGARTKFAEELRAKALELERKLTELTQLYKSLQQKVRSRARGVGKDLTGTCETEAYWKELREHVWMWLWLCVCLCVCVYVQFT

Specifications

Accession Number NP_085144.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80830
Molecular Weight (g/mol) 64.5kDa
Name APOL6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APOL-VI/APOLVI/DKFZp667M075/FLJ38562/FLJ90164/MGC57495
Common Name APOL6
Gene Symbol APOL6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.