Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Apolipoprotein A-I Quantikine ELISA Kit
SDP

Catalog No. DAPA10 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DAPA10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DAPA10 Supplier R&D Systems™ Supplier No. DAPA10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Apolipoprotein A-I Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Apolipoprotein A-I Quantikine ELISA Kit quantifies human Apolipoprotein A-I/ApoA1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (25 ul).

Specifications

Assay Range 6.25 - 200ng/mL
Assay Sensitivity 0.603ng/mL
Detectable Analytes Apolipoprotein A-I
Assay Solid Phase ELISA
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules; <50% cross-species reactivity observed with species tested
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human ApoA1
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 335, 11806, 25081
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Alp-1, APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, Apolipoprotein AI, Brp-14, Ltw-1, Lvtw-1, MGC117399, Sep-1, Sep-2
Total Assay Time 3.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.