Learn More
ACROBiosystems Human APP 2Vials
Supplier:  ACROBiosystems CHEKATP0812VIALS
Encompass_Preferred
Amyloid precursor protein (APP) is a type I integral membrane protein ubiquitously expressed in many tissues and concentrated in the synapses of neurons. It has three predominant splice variants: APP695 APP751 and APP770. The majority of APP is cleaved at the plasma membrane by the α-secretase in the non-amyloidogenic pathway. The amyloidogenic pathway starts with β-secretase cleavage by BACE1 on the N-terminal part of the Aβ domain releasing sAPPβ from a membrane-anchored fragment named βCTF or C99 which is subsequently cleaved by γ-secretase to release Aβ.
Catalog No. 50-253-5289
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.