Learn More
Human APP/Protease Nexin II (Research Grade Solanezumab Biosimilar) Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | APP |
| Applications | CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Clone | Hu13 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. |
| Gene Alias | Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II |
| Host Species | Human |
| Immunogen | Human APP/Protease Nexin II |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.