Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ARHGAP18 Synthetic Peptide

Catalog No. PEP1007
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1007 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1007 Supplier Invitrogen™ Supplier No. PEP1007
Add to Cart
Add to Cart

Synthetic Peptide

ARHGAP18 Synthetic Peptide, PEP-1007, from Invitrogen.

ARHGAP18 is one member of the human RhoGAP family with approximately 80 RhoGAP proteins known to be encoded in the human genome. Rho proteins belong to the Ras superfamily that is composed of over 50 members divided into six families, including Ras, Sar, Rho, Ran, Rab and Arf. Rho GTPases are important regulators of the actin cytoskeleton and consequently influence the shape and migration of cells. ARHGAP18 is linked to Ras, and thus, to EGFR-mediated proliferation, migration and differentiation. ARHGAP18 is precisely contained within chromosome 6q22-24, which has been shown to be linked to schizophrenia, suggesting that ARHGEP18 may play a role in this condition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 93663
Species Human
Purification Method purified
Accession Number Q8N392
Common Name Human ARHGAP18
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ARHGAP18
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.