Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARHGEF12 Partial ORF (NP_056128, 817 a.a. - 916 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89935534 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-534 25 μg
89-935-533 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-935-534 Supplier Abnova™ Supplier No. H00023365Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein is observed to form myeloid/lymphoid fusion partner in acute myeloid leukemia. [provided by RefSeq]

Sequence: VSREGILSPSELRKIFSNLEDILQLHIGLNEQMKAVRKRNETSVIDQIGEDLLTWFSGPGEEKLKHAAATFCSNQPFALEMIKSRQKKDSRFQTFVQDAE

Specifications

Accession Number NP_056128
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23365
Molecular Weight (g/mol) 36.74kDa
Name ARHGEF12 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen VSREGILSPSELRKIFSNLEDILQLHIGLNEQMKAVRKRNETSVIDQIGEDLLTWFSGPGEEKLKHAAATFCSNQPFALEMIKSRQKKDSRFQTFVQDAE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686O2372/KIAA0382/LARG/PRO2792
Common Name ARHGEF12
Gene Symbol ARHGEF12
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.