Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARL6IP5 Partial ORF (NP_006398, 1 a.a. - 64 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89935616 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-616 10 μg
89-935-617 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-935-616 Supplier Abnova™ Supplier No. H00010550Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Expression of this gene is affected by vitamin A. The encoded protein of this gene may be associated with the cytoskeleton. A similar protein in rats may play a role in the regulation of cell differentiation. The rat protein binds and inhibits the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport. [provided by RefSeq]

Sequence: MDVNIAPLRAWDDFFPGSDRFARPDFRDISKWNNRVVSNLLYYQTNYLVVAAMMISIVGFLSPF

Specifications

Accession Number NP_006398
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10550
Molecular Weight (g/mol) 32.78kDa
Name ARL6IP5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MDVNIAPLRAWDDFFPGSDRFARPDFRDISKWNNRVVSNLLYYQTNYLVVAAMMISIVGFLSPF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DERP11/GTRAP3-18/HSPC127/JWA/PRAF3/addicsin/hp22/jmx
Common Name ARL6IP5
Gene Symbol ARL6IP5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.