Learn More
Sino Biological Argininosuccinate lyase Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier: Sino Biological 200445T44100
Produced in rabbits immunized with E. coli-derived Human Argininosuccinate lyase fragment, and purified by antigen affinity chromatography.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.