Learn More
iXCells Biotechnologies Human Astrocytes (HA)
Supplier:  iXCells Biotechnologies 10HU035
Encompass_Preferred
iXCells Biotechnologies provides high quality Human Astrocytes (HA), which are isolated from human brain (cerebral cortex) and cryopreserved at P2, with >0.5 million cells in each vial. These HA express GFAP. They are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi. HA can further expand in Astrocyte Medium under the condition suggested by iXCells Biotechnologies.
Catalog No. 50-107-8008
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.