Learn More
Abcam Human ATM (phospho S1981) peptide, 1MG
Supplier:  Abcam AB950371MG
Encompass_Preferred
Human ATM (phospho S1981) peptide is a Synthetic blocking peptide. >70% purity and suitable for BL.
Protein/ Peptide/ Enzyme Synonyms: Serine-protein kinase ATM, Ataxia telangiectasia mutated, A-T mutated, ATM
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-8538