Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATP1A3 Full-length ORF (NP_689509.1, 1 a.a. - 1013 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89935995 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-995 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-995 Supplier Abnova™ Supplier No. H00000478P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes an alpha 3 subunit. [provided by RefSeq]

Sequence: MGDKKDDKDSPKKNKGKERRDLDDLKKEVAMTEHKMSVEEVCRKYNTDCVQGLTHSKAQEILARDGPNALTPPPTTPEWVKFCRQLFGGFSILLWIGAILCFLAYGIQAGTEDDPSGDNLYLGIVLAAVVIITGCFSYYQEAKSSKIMESFKNMVPQQALVIREGEKMQVNAEEVVVGDLVEIKGGDRVPADLRIISAHGCKVDNSSLTGESEPQTRSPDCTHDNPLETRNITFFSTNCVEGTARGVVVATGDRTVMGRIATLASGLEVGKTPIAIEIEHFIQLITGVAVFLGVSFFILSLILGYTWLEAVIFLIGIIVANVPEGLLATVTVCLTLTAKRMARKNCLVKNLEAVETLGSTSTICSDKTGTLTQNRMTVAHMWFDNQIHEADTTEDQSGTSFDKSSHTWVALSHIAGLCNRAVFKGGQDNIPVLKRDVAGDASESALLKCIELSSGSVKLMRERNKKVAEIPFNSTNKYQLSIHETEDPNDNRYLLVMKGAPERILDRCSTILLQGKEQPLDEEMKEAFQNAYLELGGLGERVLGFCHYYLPEEQFPKGFAFDCDDVNFTTDNLCFVGLMSMIDPPRAAVPDAVGKCRSAGIKVIMVTGDHPITAKAIAKGVGIISEGNETVEDIAARLNIPVSQVNPRDAKACVIHGTDLKDFTSEQIDEILQNHTEIVFARTSPQQKLIIVEGCQRQGAIVAVTGDGVNDSPALKKADIGVAMGIAGSDVSKQAADMILLDDNFASIVTGVEEGRLIFDNLKKSIAYTLTSNIPEITPFLLFIMANIPLPLGTITILCIDLGTDMVPAISLAYEAAESDIMKRQPRNPRTDKLVNERLISMAYGQIGMIQALGGFFSYFVILAENGFLPGNLVGIRLNWDDRTVNDLEDSYGQQWTYEQRKVVEFTCHTAFFVSIVVVQWADLIICKTRRNSVFQQGMKNKILIFGLFEETALAAFLSYCPGMDVALRMYPLKPSWWFCAFPYSFLIFVYDEIRKLILRRNPGGWVEKETYY

Specifications

Accession Number NP_689509.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 478
Molecular Weight (g/mol) 138.1kDa
Name ATP1A3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DYT12/MGC13276/RDP
Common Name ATP1A3
Gene Symbol ATP1A3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.