Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATP6V0A1 Full-length ORF (NP_005168.2, 1 a.a. - 831 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89936052 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-052 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-936-052 Supplier Abnova™ Supplier No. H00000535P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes one of three A subunit proteins and the encoded protein is associated with clathrin-coated vesicles. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGELFRSEEMTLAQLFLQSEAAYCCVSELGELGKVQFRDLNPDVNVFQRKFVNEVRRCEEMDRKLRFVEKEIRKANIPIMDTGENPEVPFPRDMIDLEANFEKIENELKEINTNQEALKRNFLELTELKFILRKTQQFFDEMADPDLLEESSSLLEPSEMGRGTPLRLGFVAGVINRERIPTFERMLWRVCRGNVFLRQAEIENPLEDPVTGDYVHKSVFIIFFQGDQLKNRVKKICEGFRASLYPCPETPQERKEMASGVNTRIDDLQMVLNQTEDHRQRVLQAAAKNIRVWFIKVRKMKAIYHTLNLCNIDVTQKCLIAEVWCPVTDLDSIQFALRRGTEHSGSTVPSILNRMQTNQTPPTYNKTNKFTYGFQNIVDAYGIGTYREINPAPYTIITFPFLFAVMFGDFGHGILMTLFAVWMVLRESRILSQKNENEMFSTVFSGRYIILLMGVFSMYTGLIYNDCFSKSLNIFGSSWSVRPMFTYNWTEETLRGNPVLQLNPALPGVFGGPYPFGIDPIWNIATNKLTFLNSFKMKMSVILGIIHMLFGVSLSLFNHIYFKKPLNIYFGFIPEIIFMTSLFGYLVILIFYKWTAYDAHTSENAPSLLIHFINMFLFSYPESGYSMLYSGQKGIQCFLVVVALLCVPWMLLFKPLVLRRQYLRRKHLGTLNFGGIRVGNGPTEEDAEIIQHDQLSTHSEDADEFDFGDTMVHQAIHTIEYCLGCISNTASYLRLWALSLAHAQLSEVLWTMVIHIGLSVKSLAGGLVLFFFFTAFATLTVAILLIMEGLSAFLHALRLHWVEFQNKFYSGTGFKFLPFSFEHIREGKFEE

Specifications

Accession Number NP_005168.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 535
Molecular Weight (g/mol) 122.2kDa
Name ATP6V0A1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ATP6N1/ATP6N1A/DKFZp781J1951/Stv1/VPP1/Vph1/a1
Common Name ATP6V0A1
Gene Symbol ATP6V0A1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.