Learn More
Description
Specifications
Specifications
| Assay Range | 31.30 to 2000 pg/mL |
| Assay Sensitivity | 4.6 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Cerebrospinal Fluid, Plasma (EDTA, Heparin), Saliva, Serum, Urine |
| Specificity | Natural and Recombinant Human Axl |
| Gene ID (Entrez) | 558 |
| Host Species | Human |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50 μL),Cell Lysates (50 μL),Serum (10 μL),EDTA Plasma (10 μL),Heparin Plasma (10 μL),Saliva (50 μL),Urine (10 μL),Cerebrospinal Fluid (10 μL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
