Learn More
Description
- Also known as: B4GALT; GALGT2
- B4GALNT2 catalyzes the last step in the biosynthesis of the human Sd(a) antigen through the addition of an N-acetylgalactosamine residue via a beta-1,4 linkage to a subterminal galactose residue substituted with an alpha-2,3-linked sialic acid.
- B4GALNT2 also catalyzes the last step in the biosynthesis of the Cad antigen.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | B4GALNT2 |
| Accession Number | NP_703147 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1714 bp |
| Gene ID (Entrez) | 124872 |
| Gene Alias | 4 GalNAcT/Sda-GalNAcT, B4GALT, beta 1,4 N-acetylgalactosaminyltransferase/betal, beta-1,4 N-acetylgalactosaminyltransferase 2, beta-1,4-N-acetyl-galactosaminyl transferase 2, beta-4-N-acetylgalactosaminyltransferase, GALGT2, MGC142235, MGC142237, sd(a) beta-1,4-GalNAc transferase, sda beta-1,4-GalNAc transferase, UDP-GalNAc:Neu5Aca2-3 Galb-R b1,4-N-acetylgalactosaminyltransferase |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
