Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human B4GALT3 Full-length ORF (NP_003770.1, 1 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89936286 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-286 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-936-286 Supplier Abnova™ Supplier No. H00008703P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. This gene encodes an enzyme that may be mainly involved in the synthesis of the first N-acetyllactosamine unit of poly-N-acetyllactosamine chains. [provided by RefSeq]

Sequence: MLRRLLERPCTLALLVGSQLAVMMYLSLGGFRSLSALFGRDQGPTFDYSHPRDVYSNLSHLPGAPGGPPAPQGLPYCPERSPLLVGPVSVSFSPVPSLAEIVERNPRVEPGGRYRPAGCEPRSRTAIIVPHRAREHHLRLLLYHLHPFLQRQQLAYGIYVIHQAGNGTFNRAKLLNVGVREALRDEEWDCLFLHDVDLLPENDHNLYVCDPRGPRHVAVAMNKFGYSLPYPQYFGGVSALTPDQYLKMNGFPNEYWGWGGEDDDIATRVRLAGMKISRPPTSVGHYKMVKHRGDKGNEENPHRFDLLVRTQNSWTQDGMNSLTYQLLARELGPLYTNITADIGTDPRGPRAPSGPRYPPGSSQAFRQEMLQRRPPARPGPLSTANHTALRGSH

Specifications

Accession Number NP_003770.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8703
Molecular Weight (g/mol) 70.3kDa
Name B4GALT3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias beta4Gal-T3
Common Name B4GALT3
Gene Symbol B4GALT3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.