Learn More
Kingfisher Biotech Inc Human B7-H2 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1710H500
Encompass_Preferred
The Human B7-H2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human B7-H2 applications are for cell culture, ELISA standard, and Western Blot Control. Human B7-H2 yeast-derived recombinant protein can be purchased in multiple sizes. Human B7-H2 Specifications: (Molecular Weight: 26.4 kDa) (Amino Acid Sequence: DTQEKEVRAM VGSDVELSCA CPEGSRFDLN DVYVYWQTSE SKTVVTYHIP QNSSLENVDS RYRNRALMSP AGMLRGDFSL RLFNVTPQDE QKFHCLVLSQ SLGFQEVLSV EVTLHVAANF SVPVVSAPHS PSQDELTFTC TSINGYPRPN VYWINKTDNS LLDQALQNDT VFLNMRGLYD VVSVLRIART PSVNIGCCIE NVLLQQNLTV GSQTGNDIGE RDKITENPVS TGEKNAAT (238)) (Gene ID: 23308). For research use only. Made in the USA
Catalog No. 50-253-2909
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.