Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human B7-H3 Quantikine ELISA Kit
SDP

Catalog No. DB7H30 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DB7H30 1 Kit
SB7H30 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DB7H30 Supplier R&D Systems™ Supplier No. DB7H30

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human B7-H3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human B7-H3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human B7-H3/CD276 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (50 ul).

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.274 ng/mL
Conjugate HRP
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Plasma (EDTA, Heparin), Serum, Serum, Urine, Urine
Specificity Natural and Recombinant Human B7-H3
For Use With (Application) ELISA
Gene ID (Entrez) 80381
Gene Symbol CD276
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule
Target B7-H3/CD276
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.