Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit

Catalog No. SBLYS0B
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SBLYS0B 6 Plates 6 x 96-well Plates
DBLYS0B 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SBLYS0B Supplier R&D Systems™ Supplier No. SBLYS0B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit quantifies human BAFF/BLyS/TNFSF13B in cell culture supernates (25 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 6.44pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human BAFF/BLyS
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 10673
Quantity 6 Plates
Sample Volume Cell Culture Supernates (25uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1 delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13 b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13 B, tumor necrosis factor superfamily, member 13 b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.