Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BAFF Receptor Recombinant Protein, PeproTech®

Catalog No. 31013R500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31013R500UG 500 μg
31013R100UG 100 μg
31013R10UG 10 μg
310-13R-1MG 1 mg
31013R250UG 250 μg
31013R50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 31013R500UG Supplier Gibco™ Supplier No. 31013R500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BAFFR is a 76 amino acid polypeptide (7.7 kDa) corresponding to the extracellular portion of the full BAFFR protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96RJ3
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 115650
Molecular Weight (g/mol) 7.7 kDa
Name Human BAFF Receptor
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13 C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13 C; tumor necrosis factor receptor superfamily, member 13 C
Common Name BAFF Receptor (CD268)
Gene Symbol TNFRSF13C
Biological Activity Determined by its ability to block BAFF induced mouse splenocyte survival. The expected ED50 for this effect is 2.0-4.0 ug/ml in the presence of 1.0 ug/ml of human soluble BAFF.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MRRGPRSLRG RDAPAPTPCV PAECFDLLVR HCVACGLLRT PRPKPAGASS PAPRTALQPQ ESVGAGAGEA ALPLPG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.