Learn More
Gemini Bio Products HUMAN BASIC FIBROBLAST GF 50UG
SDP
Supplier:  Gemini Bio Products 300112P50UG
Encompass_Preferred
Basic fibroblast growth factor is also known as bFGF, FGF2 or FGF-beta. It is used to support the culture of embryonic stem cells. This carrier-free product is a recombinant human protein expressed in E. coli.
Catalog No. NC0584791
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.