Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BCAS1 Full-length ORF (ADR83077.1, 1 a.a. - 584 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89936479 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-479 25 μg
89-936-478 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-936-479 Supplier Abnova™ Supplier No. H00008537P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene resides in a region at 20q13 which is amplified in a variety of tumor types and associated with more aggressive tumor phenotypes. Among the genes identified from this region, it was found to be highly expressed in three amplified breast cancer cell lines and in one breast tumor without amplification at 20q13.2. However, this gene is not in the common region of maximal amplification and its expression was not detected in the breast cancer cell line MCF7, in which this region is highly amplified. Although not consistently expressed, this gene is a candidate oncogene. [provided by RefSeq]

Sequence: MGNQMSVPQRVEDQENEPEAETYKDNASALNGVPVVVSTHTVQHLEEVDLGISVKTDNVATSSPETTEISAVADANGKNLGKEAKPEAPAAKSRFFLMLSRPVPGRTGDQAADSSLGSVKLDVSSNKAPANKDPSESWTLPVAAGPGQDTDKTPGHAPAQDKALSAARDPTLLPPETGGAGGEAPSKPKDSSFFDKFFKLDKGQEKVPGDSQQEAKRAEHQDKVDEVPGLSGQSDDVPAGKDIVDGKEKEGQELGTADCSVPGDPEGLETAKDDSQAAAIAENNNSIMSFFKTLVSPNKAETKKDPEDTGAEKSPTTSADLKSDKANFTSQETQGAGKNSKGCNPSGHTQSVTTPEPAKEGTKEKSGPTSLPLGKLFWKKSVKEDSVPTGAEENVVCESPVEIIKSKEVESALQTVDLNEGDAAPEPTEAKLKREESKPRTSLMAFLRQMSVKGDGGITHSEEINGKDSSCQTSDSTEKTITPPEPEPTGAPQKGKEGSSKDKKSAAEMNKQKSNKQEAKEPAQCTEQATVDTNSLQNGDKLQKRPEKRQQSLGGFFKGLGPKRMLDAQVQTDPVSIGPVGKSK

Specifications

Accession Number ADR83077.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8537
Molecular Weight (g/mol) 90.7kDa
Name BCAS1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AIBC1/NABC1
Common Name BCAS1
Gene Symbol BCAS1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.