Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BCL6 (P41182, 1 a.a. - 150 a.a.) Partial Recombinant Protein

Catalog No. 89936541 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-541 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-936-541 Supplier Abnova™ Supplier No. P4858

Used for SDS-PAGE

The protein encoded by this gene is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors. This gene is found to be frequently translocated and hypermutated in diffuse large-cell lymphoma (DLCL), and may be involved in the pathogenesis of DLCL. Alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq]

Specifications

Accession Number P41182
Concentration 0.1mg/mL
For Use With (Application) SDS-PAGE
Formulation In 50mM Tris-HCl, 10mM GSH, pH 7.5
Gene ID (Entrez) 604
Molecular Weight (g/mol) 43kDa
Name Human BCL6 (P41182, 1 a.a. - 150 a.a.) Partial Recombinant Protein
Quantity 10 μg
Storage Requirements Store at −80°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Common Name BCL6
Gene Symbol BCL6
Species E. coli
Recombinant Recombinant
Protein Tag Untagged
Expression System Escherichia coli expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.