Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BCL9 Partial ORF (NP_004317, 1036 a.a. - 1135 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89936559 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-559 10 ug
89-936-560 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-936-559 Supplier Abnova™ Supplier No. H00000607Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

BCL9 is associated with B-cell acute lymphoblastic leukemia. It may be a target of translocation in B-cell malignancies with abnormalities of 1q21. Its function is unknown. The overexpression of BCL9 may be of pathogenic significance in B-cell malignancies. [provided by RefSeq]

Sequence: DDDSPPARSPNLPSMNNMPGMGINTQNPRISGPNPVVPMPTLSPMGMTQPLSHSNQMPSPNAVGPNIPPHGVPMGPGLMSHNPIMGHGSQEPPMVPQGRM

Specifications

Accession Number NP_004317
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 607
Molecular Weight (g/mol) 36.74kDa
Name BCL9 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen DDDSPPARSPNLPSMNNMPGMGINTQNPRISGPNPVVPMPTLSPMGMTQPLSHSNQMPSPNAVGPNIPPHGVPMGPGLMSHNPIMGHGSQEPPMVPQGRM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LGS/MGC131591
Common Name BCL9
Gene Symbol BCL9
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.