Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BCMA Recombinant Protein, PeproTech®

Catalog No. 31016100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31016100UG 100 μg
310-16-1MG 1 mg
310-16-20UG 20 μg
31016250UG 250 μg
31016500UG 500 μg
310-16-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 31016100UG missing translation for 'mfr' Gibco™ missing translation for 'supplierNo' 31016100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant soluble Human BCMA is a 50 amino acid polypeptide (5.3 kDa) comprising the TNFR homologous region of the BCMA protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is preferentially expressed in mature B lymphocytes, and may be important for B cell development and autoimmune response. This receptor has been shown to specifically bind to the tumor necrosis factor (ligand) superfamily, member 13b (TNFSF13B/TALL-1/BAFF), and to lead to NF-kappaB and MAPK8/JNK activation. This receptor also binds to various TRAF family members, and thus may transduce signals for cell survival and proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02223
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 608
Molecular Weight (g/mol) 5.3 kDa
Name Human BCMA
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias B cell maturation antigen; B-cell maturation factor; B-cell maturation protein; BCM; BCMA; CD269; TNF receptor superfamily member 17; Tnfrsf13; Tnfrsf13a; Tnfrsf17; Tumor necrosis factor receptor superfamily member 17; tumor necrosis factor receptor superfamily, member 17
Common Name BCMA
Gene Symbol Tnfrsf17
Biological Activity Determined by its ability to neutralize APRIL-induced NF-?B activation in HEK293-NF-?B reporter cells transfected with BCMA receptor.
Conjugate Unconjugated
Recombinant Recombinant
Sequence AGQCSQNEYF DSLLHACIPC QLRCSSNTPP LTCQRYCNAS VTNSVKGTNA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.