Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human BCMA/TNFRSF17 Quantikine ELISA Kit

Catalog No. DBCMA0
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DBCMA0 1 Kit
SBCMA0 1 Pack (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DBCMA0 Supplier R&D Systems™ Supplier No. DBCMA0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 10.8 pg/mL
Sample Type Cell Culture Supernatant, Serum, Plasma (EDTA), Heparin Plasma, Saliva, Urine
Specificity Natural and recombinant human BCMA.
For Use With (Application) ELISA
Gene ID (Entrez) 608
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.