Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human BCMA/TNFRSF17 Quantikine ELISA Kit
SDP

Catalog No. SBCMA0 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SBCMA0 1 Pack (6 Plates)
DBCMA0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SBCMA0 Supplier R&D Systems™ Supplier No. SBCMA0

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 10.8 pg/mL
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum, Plasma (EDTA), Heparin Plasma, Saliva, Urine
Specificity Natural and recombinant human BCMA.
For Use With (Application) ELISA
Gene ID (Entrez) 608
Quantity 1 Pack (6 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.