Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BD-2 Recombinant Protein, PeproTech®

Catalog No. 300495UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-49-5UG 5 μg
30049100UG 100 μg
300-49-1MG 1 mg
300-49-20UG 20 μg
30049250UG 250 μg
30049500UG 500 μg
300-49-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 300-49-5UG Supplier Gibco™ Supplier No. 300495UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BD-2 is a 4.3 kDa protein containing 41 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15263
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 1673
Molecular Weight (g/mol) 4.3 kDa
Name Human BD-2
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BD-2; beta defensin 2; beta defensin-2; Beta-defensin 2; Beta-defensin 4 A; DEFB102; DEFB2; DEFB-2; DEFB4; DEFB4A; DEFB4B; Defensin beta 2; defensin beta 4 A; Defensin beta2; defensin, beta 2; defensin, beta 4; defensin, beta 4 A; Defensin-beta2; HBD-2; SAP1; Skin-antimicrobial peptide 1
Common Name beta-2 Defensin
Gene Symbol DEFB4A
Biological Activity Determined by its ability to chemoattract immature human dendritic cells using a concentration of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence GIGDPVTCLK SGAICHPVFC PRRYKQIGTC GLPGTKCCKK P
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.