Learn More
Abcam Human BD-3 Synthetic Peptide, >95% Purity, 100 μg
Description
- Peptide Type: Synthetic
- Purity: >95%
- Form: Lyophilized in 0.1% Acetic acid
- Tested Applications: BL
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Beta-Defensin 3 (Human) |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Buffer | 0.1% acetic acid |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Cross Reactivity | Human |
| Formulation | Lyophilized from 0.1% Acetic acid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.