Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BD-4 Recombinant Protein, PeproTech®

Catalog No. 3006520UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-65-20UG 20 μg
30065100UG 100 μg
300-65-1MG 1 mg
30065250UG 250 μg
30065500UG 500 μg
300-65-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 300-65-20UG Supplier Gibco™ Supplier No. 3006520UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BD-4 is a 6.0 kDa protein containing 50 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WTQ1
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 140596
Molecular Weight (g/mol) 6 kDa
Name Human BD-4
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BD-4; Beta-defensin 104; Beta-defensin 4; DEFB104; DEFB104A; DEFB104B; DEFB4; DEFB-4; defensin beta 104 A; Defensin, beta 104; defensin, beta 104 A; defensin, beta 4; hBD-4
Common Name DEFB104A
Gene Symbol DEFB104A
Biological Activity Determined by its ability to chemoattract human monocytes using a concentration range of 0.1-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.