Learn More
Kamiya Biomedical HUMAN BETA 2-MICROGLOBULIN (B2M) ELISA, 96 Tests
SDP
Supplier:  Kamiya Biomedical KT500
Encompass
HUMAN BETA 2-MICROGLOBULIN (B2M) ELISA, 96 Tests. For the quantitative determination of B2M in human biological samples. Assay range: 3.13 - 100 ng/mL. Includes calibrator and control.
Catalog No. NC1601112
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.