Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human beta-Defensin 1 (36 aa) Recombinant Protein, PeproTech®

Catalog No. 3005120UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-51-20UG 20 μg
30051100UG 100 μg
300-51-1MG 1 mg
30051250UG 250 μg
30051500UG 500 μg
300-51-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 300-51-20UG Supplier Gibco™ Supplier No. 3005120UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BD-1 is a 3.9 kDa protein containing 36 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60022
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 1672
Molecular Weight (g/mol) 3.9 kDa
Name Human beta-Defensin 1 (36 aa)
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AW260221; BD1; BD-1; beta defensin 1; beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; defensin, beta 1; HBD1; rBD-1
Common Name beta-Defensin 1
Gene Symbol DEFB1
Biological Activity Determined by its ability to chemoattract CD34+ dendritic cells using a concentration range of 100.0-1000.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence DHYNCVSSGG QCLYSACPIF TKIQGTCYRG KAKCCK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.