Learn More
Raybiotech Inc HUMAN BETA-NGF ELISA
Supplier:  Raybiotech Inc ELHBNGF1
Encompass_Preferred
RayBio Human beta-NGF ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 14 pg/ml - 5000 pg/ml
Catalog No. 50-104-3509
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.