Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Bikunin DuoSet ELISA
SDP

Catalog No. DY774405 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
Five 96-well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DY774405 Five 96-well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY774405 Supplier R&D Systems™ Supplier No. DY774405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Bikunin.

The Human Bikunin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Bikunin.

Specifications

Assay Range 6.25 to 400pg/mL
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sufficient For Five 96-well plates
For Use With (Application) ELISA
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.