Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Bladder Tissue MicroArray (Cancer)
SDP

Catalog No. NB397860 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397860 1 Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397860 Supplier R&D Systems™ Supplier No. NBP311846

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. No. of samples: 59

Specifications

Quantity 1 Slide
Species Human
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Paraffin
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.