Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BLK (NM_001715.2, 1 a.a. - 505 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells

Catalog No. 89936683 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-936-683 Supplier Abnova™ Supplier No. P4635
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Specifications

Accession Number NM_001715.2
Concentration 0.120 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 50mM Tris-HCl, 100mM NaCl, pH 8.0. (5mM DTT, 4mM reduced glutathione, 20% glycerol)
Gene ID (Entrez) 640
Molecular Weight (g/mol) 87.102kDa
Name BLK (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method One-step affinity purification using GSH-agarose
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias MGC10442
Common Name BLK
Gene Symbol BLK
Biological Activity 167pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST-His
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.