Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BLNK Full-length ORF (AAH18906.1, 1 a.a. - 456 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89936694 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-694 10 μg
89-936-695 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-936-694 Supplier Abnova™ Supplier No. H00029760P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a cytoplasmic linker or adaptor protein that plays a critical role in B cell development. This protein bridges B cell receptor-associated kinase activation with downstream signaling pathways, thereby affecting various biological functions. The phosphorylation of five tyrosine residues is necessary for this protein to nucleate distinct signaling effectors following B cell receptor activation. Mutations in this gene cause hypoglobulinemia and absent B cells, a disease in which the pro- to pre-B-cell transition is developmentally blocked. Deficiency in this protein has also been shown in some cases of pre-B acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MDKLNKITVPASQKLRQLQKMVHDIKNNEGGIMNKIKKLKVKAPPSVPRRDYASESPADEEQQWSDDFDSDYENPDEHSDSEMYVMPAEENADDSYEPPPVEQETRPVHPALPFARGEYIDNRSSQRHSPPFSKTLPSKPSWPSEKARLTSTLPALTALQKPQVPPKPKGLLEDEADYVVPVEDNDENYIHPTESSSPPPEKAPMVNRSTKPNSSTPASPPGTASGRNSGAWETKSPPPAAPSPLPRAGKKPTTPLKTTPVASQQNASSVCEEKPIPAERHRGSSHRQEAVQSPVFPPAQKQIHQKPIPLPRFTEGGNPTVDGPLPSFSSNSTISEQEAGVLCKPWYAGACDRKSAEEALHRSNKDGSFLIRKSSGHDSKQPYTLVVFFNKRVYNIPVRFIEATKQYALGRKKNGEEYFGSVAEIIRNHQHSPLVLIDSQNNTKDSTRLKYAVKVS

Specifications

Accession Number AAH18906.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 29760
Molecular Weight (g/mol) 76.9kDa
Name BLNK (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BASH/BLNK-S/LY57/MGC111051/SLP-65/SLP65
Common Name BLNK
Gene Symbol BLNK
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.