Learn More
Description
- Also known as: PCP; TLD; OI13; PCP2; PCOLC
- BMP-1 is a zinc protease of the astacin family.
- BMP-1 plays a key role in formation of extracellular matrix (ECM) by converting precursor proteins into their mature and functional forms.
- The precursor proteins identified as substrates for BMP-1 include collagens, biglycan, laminin 5, dentin matrix protein-1, and lysyl oxidase.
- Alternatively spliced transcripts encoding different proteins have been described.
- The BMP-1/PCP insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | BMP-1/PCP |
| Accession Number | NP_006120 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2974 bp |
| Gene ID (Entrez) | 649 |
| Gene Alias | BMP1, BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
