Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BMP-10 Recombinant Protein, PeproTech®

Catalog No. 12040100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12040100UG 100 μg
120-40-10UG 10 μg
120-40-1MG 1 mg
12040500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 12040100UG Supplier Gibco™ Supplier No. 12040100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BMP-10 is a 24.4 kDa homodimeric disulfide-linked protein consisting of two 108 amino acid subunits, which correspond to amino acid residues 317 to 424 of the full-length BMP-10 precursor. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body and are involved in diverse physiological processes during both pre- and postnatal life. BMP-10 plays a crucial role in the development of the embryonic heart by acting to stimulate and maintain cardiomyocyte proliferation. It can signal through various receptor complexes usually containing BMPR-1A, BMPR-1B, ALK1, ALK3, or ALK6. The interaction of BMP-10 with its specific receptors can induce signaling initiated by the phosphorylation of SMAD transcription factors, including SMAD1, SMAD5, or SMAD8, but can also induce SMAD independent processes. BMP-10 is structurally related to BMP-9, and both can inhibit endothelial cell proliferation and migration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95393
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27302
Molecular Weight (g/mol) 24.4 kDa
Name Human BMP-10
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BMP; BMP10; BMP-10; bmp10 {ECO:0000312; bmp10b; Bone morphogenetic protein; bone morphogenetic protein 10; EMBL:BAE16990.1, ECO:0000312; RGD:1562986}; si:dkey-13n15.5
Common Name BMP-10
Gene Symbol BMP10
Biological Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 4.0-6.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence NAKGNYCKRT PLYIDFKEIG WDSWIIAPPG YEAYECRGVC NYPLAEHLTP TKHAIIQALV HLKNSQKASK ACCVPTKLEP ISILYLDKGV VTYKFKYEGM AVSECGCR
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.