Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BMP-13/CDMP-2 Recombinant Protein, PeproTech®

Catalog No. 12004250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12004250UG 250 μg
12004100UG 100 μg
120-04-10UG 10 μg
120-04-1MG 1 mg
12004500UG 500 μg
120-04-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12004250UG Supplier Gibco™ Supplier No. 12004250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BMP-13/CDMP-2 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF6 is a growth differentiation factor (GDFs), which are members of the transforming growth factor (TGF) superfamily that is involved in embryonic development and adult tissue homeostasis. Both GDF6 and GDF7 are closely related to GDF5 which has been shown to induce activation of plasminogen activator, thereby inducing angiogenesis. It is predominantly expressed in long bones during fetal embryonic development and is involved in bone formation. In Xenopus, GDF6 is expressed at the edge of the neural plate and within the anterior neural plate including the eye fields. GDF6 is required for normal formation of some bones and joints in the limbs, skull, and axial skeleton. It may regulate patterning of the ectoderm by interacting with bone morphogenetic proteins (BMPs), and control eye development. Mutations in this gene result in colobomata, which are congenital abnormalities in ocular development, and in Klippel-Feil syndrome (KFS), a congenital disorder of spinal segmentation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6KF10
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 392255
Molecular Weight (g/mol) 27 kDa
Name Human BMP-13/CDMP-2
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BMP13; BMP-13; Bone morphogenetic protein 13; CDMP2; GDF16; GDF6; Gdf-6; growth differentiation factor 16; growth differentiation factor 6; growth/differentiation factor 16; Growth/differentiation factor 6; KFM; KFS; KFS1; KFSL; Klip-Feil malformation; Klippel-Feil malformation; Klippel-Feil syndrome; LCA17; MCOP4; MCOPCB6; SCDO4; SGM1
Common Name GDF6
Gene Symbol GDF6
Biological Activity The ED50 was determined by its ability to induce alkaline phosphatase production by ATDC-5 chondrogenic cells in the range of 2.0-3.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence TAFASRHGKR HGKKSRLRCS KKPLHVNFKE LGWDDWIIAP LEYEAYHCEG VCDFPLRSHL EPTNHAIIQT LMNSMDPGST PPSCCVPTKL TPISILYIDA GNNVVYKQYE DMVVESCGCR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.