Learn More
Invitrogen™ Human BMP-15 ELISA Kit
Description
Human BMP-15 quantitates human BMP-15 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BMP-15.
Specifications
Specifications
| Accession Number | AU015375; AU018861; AU021453; BMP; Bmp15; Bmp-15; Bone morphogenetic protein; bone morphogenetic protein 15; C86824; C87336; GDF9B; Gdf-9 b; growth differentiation factor-9 B; growth/differentiation factor 9 B; ODG2; POF4 |
| Assay Range | 0.6 to 150 ng/mL |
| Assay Sensitivity | 0.6 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 9210 |
| Gene Symbol | BMP15 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.